accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOOBr

CAS RN 37742-98-6

IUPAC Name: 4-bromo-2,2-diphenylbutanoic acid
Synonyms: 4-bromo-2,2-diphenylbutanoic acid
Molecular Weight (g/mol): 319.2
Molecular Formula: C16H15BrO2
InChi Key: GFIYIIRFIODLLU-UHFFFAOYSA-N
SMILES: OC(=O)C(CCBr)(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results