CAS RN 3777-70-6
IUPAC Name:
2-hexylfuran
Synonyms:
2-hexylfuran
Molecular Weight (g/mol):
152.24
Molecular Formula:
C10H16O
InChi Key:
XBLCAKKYMZVLPU-UHFFFAOYSA-N
SMILES:
CCCCCCC1=CC=CO1
1
–
1
of
1
results
1
2-n-Hexylfuran, 97%
CAS: 3777-70-6 Molecular Formula: C10H16O Molecular Weight (g/mol): 152.237 MDL Number: MFCD00053134 InChI Key: XBLCAKKYMZVLPU-UHFFFAOYSA-N Synonym: 2-n-hexylfuran,furan, 2-hexyl,2-hexyl-furan,acmc-1clja PubChem CID: 77408 IUPAC Name: 2-hexylfuran SMILES: CCCCCCC1=CC=CO1