accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3777-71-7

IUPAC Name: 2-heptylfuran
Synonyms: 2-heptylfuran
Molecular Weight (g/mol): 166.26
Molecular Formula: C11H18O
InChi Key: BHTUFJXTYNLISA-UHFFFAOYSA-N
SMILES: CCCCCCCC1=CC=CO1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results