accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 37779-49-0

IUPAC Name: methyl 3-oxo-4-phenylbutanoate
Synonyms: methyl 3-oxo-4-phenylbutanoate
Molecular Weight (g/mol): 192.21
Molecular Formula: C11H12O3
InChi Key: DTMSEOVTDVSPDO-UHFFFAOYSA-N
SMILES: COC(=O)CC(=O)CC1=CC=CC=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results