accessibility menu, dialog, popup

UserName

Viewing profile as user:
OSS

CAS RN 3779-27-9

IUPAC Name: [2,2'-bithiophene]-5-carbaldehyde
Synonyms: [2,2'-bithiophene]-5-carbaldehyde
Molecular Weight (g/mol): 194.27
Molecular Formula: C9H6OS2
InChi Key: FYBWRAXKYXTOQC-UHFFFAOYSA-N
SMILES: O=CC1=CC=C(S1)C1=CC=CS1
Percent Purity 
  • (3)
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results