accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOOOH3C

CAS RN 3779-29-1

IUPAC Name: 1,1-diethyl cyclobutane-1,1-dicarboxylate
Synonyms: 1,1-diethyl cyclobutane-1,1-dicarboxylate
Molecular Weight (g/mol): 200.23
Molecular Formula: C10H16O4
InChi Key: JPNJEJSZSMXWSV-UHFFFAOYSA-N
SMILES: CCOC(=O)C1(CCC1)C(=O)OCC
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)