accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 378-75-6

IUPAC Name: methyl 2,2,3,3,3-pentafluoropropanoate
Synonyms: methyl 2,2,3,3,3-pentafluoropropanoate
Molecular Weight (g/mol): 178.06
Molecular Formula: C4H3F5O2
InChi Key: JMKJCPUVEMZGEC-UHFFFAOYSA-N
SMILES: COC(=O)C(F)(F)C(F)(F)F
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results