accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHO(R)(S)(S)(R)HOOOHOHOO

CAS RN 3786-91-2

IUPAC Name: (1R,2R,3S,4S)-cyclopentane-1,2,3,4-tetracarboxylic acid
Synonyms: (1R,2R,3S,4S)-cyclopentane-1,2,3,4-tetracarboxylic acid
Molecular Weight (g/mol): 246.17
Molecular Formula: C9H10O8
InChi Key: WOSVXXBNNCUXMT-VERZDPOYSA-N
SMILES: OC(=O)[C@H]1C[C@H]([C@H]([C@H]1C(O)=O)C(O)=O)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results