accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOBr

CAS RN 37865-96-6

IUPAC Name: 2-(2-bromoethyl)-2-methyl-1,3-dioxolane
Synonyms: 2-(2-bromoethyl)-2-methyl-1,3-dioxolane
Molecular Weight (g/mol): 195.06
Molecular Formula: C6H11BrO2
InChi Key: SZLPRRVCSGZARF-UHFFFAOYSA-N
SMILES: CC1(CCBr)OCCO1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results