accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 37895-73-1

IUPAC Name: 1,2-dibromo-4,5-dimethoxybenzene
Synonyms: 1,2-dibromo-4,5-dimethoxybenzene
Molecular Weight (g/mol): 295.96
Molecular Formula: C8H8Br2O2
InChi Key: ZYCLQXMMFJREPJ-UHFFFAOYSA-N
SMILES: COC1=CC(Br)=C(Br)C=C1OC
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)