CAS RN 379-50-0
IUPAC Name:
fluorotriphenylsilane
Synonyms:
fluorotriphenylsilane
Molecular Weight (g/mol):
278.4
Molecular Formula:
C18H15FSi
InChi Key:
UBGXLEFOIVWVRP-UHFFFAOYSA-N
SMILES:
F[Si](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
1
–
1
of
1
results
1
Fluorotriphenylsilane 97.0+%, TCI America™
CAS: 379-50-0 Molecular Formula: C18H15FSi Molecular Weight (g/mol): 278.401 MDL Number: MFCD00017899 InChI Key: UBGXLEFOIVWVRP-UHFFFAOYSA-N Synonym: Triphenylfluorosilane, Triphenylsilyl Fluoride PubChem CID: 67837 IUPAC Name: fluoro(triphenyl)silane SMILES: C1=CC=C(C=C1)[Si](C2=CC=CC=C2)(C3=CC=CC=C3)F