accessibility menu, dialog, popup

UserName

Viewing profile as user:
HFSnH

CAS RN 379-52-2

IUPAC Name: —
Synonyms: —
Molecular Weight (g/mol): 371.04
Molecular Formula: C18H17FSn
InChi Key: SZNZEEYJVCQHCQ-UHFFFAOYSA-N
SMILES: F.C1=CC=C(C=C1)[SnH](C1=CC=CC=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results