accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 37920-25-5

IUPAC Name: 1-(4-butylphenyl)ethan-1-one
Synonyms: 1-(4-butylphenyl)ethanone
Molecular Weight (g/mol): 176.26
Molecular Formula: C12H16O
InChi Key: MQESVSITPLILCO-UHFFFAOYSA-N
SMILES: CCCCC1=CC=C(C=C1)C(C)=O
Boiling Point 
  • (2)
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (1)
  • (3)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (1)
  • (3)
  • (1)
  • (1)
  • (4)