accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 37942-01-1

IUPAC Name: 5-bromo-2-methoxyphenol
Synonyms: 5-bromo-2-methoxyphenol
Molecular Weight (g/mol): 203.04
Molecular Formula: C7H7BrO2
InChi Key: OLSJHVZRUFFIPL-UHFFFAOYSA-N
SMILES: COC1=CC=C(Br)C=C1O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results