accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOOHOOOHOPOHOHO

CAS RN 37971-36-1

IUPAC Name: 2-phosphonobutane-1,2,4-tricarboxylic acid
Synonyms: phosphonobutanetricarboxylic acid
Molecular Weight (g/mol): 270.13
Molecular Formula: C7H11O9P
InChi Key: SZHQPBJEOCHCKM-UHFFFAOYNA-N
SMILES: OC(=O)CCC(CC(O)=O)(C(O)=O)P(O)(O)=O
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results