accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 37989-92-7

IUPAC Name: 1-{2'-fluoro-[1,1'-biphenyl]-4-yl}propan-1-one
Synonyms: 1-{2'-fluoro-[1,1'-biphenyl]-4-yl}propan-1-one
Molecular Weight (g/mol): 228.27
Molecular Formula: C15H13FO
InChi Key: ZXIQZWXVSBJPTC-UHFFFAOYSA-N
SMILES: CCC(=O)C1=CC=C(C=C1)C1=CC=CC=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results