accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOH3C

CAS RN 38002-88-9

IUPAC Name: 2,2-dimethyl-2,3-dihydro-1-benzofuran-7-carbaldehyde
Synonyms: 2,2-dimethyl-3H-1-benzofuran-7-carbaldehyde
Molecular Weight (g/mol): 176.22
Molecular Formula: C11H12O2
InChi Key: HGKYVFOYQUSRQN-UHFFFAOYSA-N
SMILES: CC1(C)CC2=CC=CC(C=O)=C2O1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results