accessibility menu, dialog, popup

UserName

Viewing profile as user:
NNNHO

CAS RN 38036-61-2

IUPAC Name: 2-(3-oxo-3,4-dihydroquinoxalin-2-yl)-2-phenylacetonitrile
Synonyms: 2-(3-oxo-4H-quinoxalin-2-yl)-2-phenylacetonitrile
Molecular Weight (g/mol): 261.28
Molecular Formula: C16H11N3O
InChi Key: JPISUJFDUFVJAO-UHFFFAOYNA-N
SMILES: O=C1NC2=CC=CC=C2N=C1C(C#N)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results