accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH3O

CAS RN 38041-19-9

IUPAC Name: oxan-4-aminium
Synonyms: oxan-4-aminium
Molecular Weight (g/mol): 102.16
Molecular Formula: C5H12NO
InChi Key: AHVQYHFYQWKUKB-UHFFFAOYSA-O
SMILES: [NH3+]C1CCOCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results