accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNHNCH3SOOHO

CAS RN 380416-14-8

IUPAC Name: methanesulfonic acid; methyl[2-(pyridin-2-yl)ethyl]amine
Synonyms: betahistine; methanesulfonic acid
Molecular Weight (g/mol): 232.3
Molecular Formula: C9H16N2O3S
InChi Key: FUWLKZQYMWTFBA-UHFFFAOYSA-N
SMILES: CS(O)(=O)=O.CNCCC1=CC=CC=N1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results