accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 380427-39-4

IUPAC Name: methyl 2-oxo-2,3-dihydro-1H-indole-7-carboxylate
Synonyms: methyl 2-oxo-1,3-dihydroindole-7-carboxylate
Molecular Weight (g/mol): 191.19
Molecular Formula: C10H9NO3
InChi Key: XVJRNLIMSQFUAI-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=CC2=C1NC(=O)C2

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results