accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 380430-55-7

IUPAC Name: hydrogen [2-amino-4-(methoxycarbonyl)phenyl]boronic acid chloride
Synonyms: hydrogen 2-amino-4-(methoxycarbonyl)phenylboronic acid chloride
Molecular Weight (g/mol): 231.44
Molecular Formula: C8H11BClNO4
InChi Key: IDUSDMZTKZZVAW-UHFFFAOYSA-N
SMILES: [H+].[Cl-].COC(=O)C1=CC=C(B(O)O)C(N)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)