accessibility menu, dialog, popup

UserName

Viewing profile as user:
SN

CAS RN 380626-35-7

IUPAC Name: 3-(thiophen-2-yl)benzonitrile
Synonyms: 3-(thiophen-2-yl)benzonitrile
Molecular Weight (g/mol): 185.24
Molecular Formula: C11H7NS
InChi Key: DIILPQRSOQWSCU-UHFFFAOYSA-N
SMILES: N#CC1=CC(=CC=C1)C1=CC=CS1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results