accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOHH2N(E/Z)

CAS RN 38090-53-8

IUPAC Name: 4-aminobut-2-enoic acid
Synonyms: (Z)-4-amino-2-butenoic acid
Molecular Weight (g/mol): 101.11
Molecular Formula: C4H7NO2
InChi Key: FMKJUUQOYOHLTF-UHFFFAOYSA-N
SMILES: NCC=CC(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results