accessibility menu, dialog, popup

UserName

Viewing profile as user:
NaONS

CAS RN 3811-73-2

IUPAC Name: sodium 2-sulfanylidene-1,2-dihydropyridin-1-olate
Synonyms: pyrithione sodium (innm) (2)
Molecular Weight (g/mol): 149.14
Molecular Formula: C5H4NNaOS
InChi Key: XNRNJIIJLOFJEK-UHFFFAOYSA-N
SMILES: [Na+].[O-]N1C=CC=CC1=S
Molecular Weight (g/mol) 
  • (8)
  • (5)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
  • (3)
  • (1)
  • (2)
Physical Form 
  • (2)
  • (1)
  • (3)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (3)
  • (8)
  • (8)
  • (5)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
  • (2)
  • (2)
  • (1)
  • (3)
  • (3)
  • (3)