accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 38111-44-3

IUPAC Name: zinc(2+) benzenide bromide
Synonyms: zinc(2+) benzenide bromide
Molecular Weight (g/mol): 222.39
Molecular Formula: C6H5BrZn
InChi Key: HMFWJMIDCMEJHO-UHFFFAOYSA-M
SMILES: [Zn++].[Br-].C1=CC=[C-]C=C1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)