CAS RN 381233-75-6
IUPAC Name:
5-bromo-3-iodo-1,2-dihydropyridin-2-one
Synonyms:
5-bromo-3-iodo-1H-pyridin-2-one
Molecular Weight (g/mol):
299.89
Molecular Formula:
C5H3BrINO
InChi Key:
WAUPPDAXWKLNNU-UHFFFAOYSA-N
SMILES:
BrC1=CNC(=O)C(I)=C1
1
–
1
of
1
results
1
Promotions Available
5-Bromo-3-iodo-2-hydroxypyridine, 97%
5-Bromo-3-iodo-2-hydroxypyridine, 97%
CAS: 381233-75-6 Molecular Formula: C5H3BrINO Molecular Weight (g/mol): 299.893 MDL Number: MFCD08688586 InChI Key: WAUPPDAXWKLNNU-UHFFFAOYSA-N PubChem CID: 12027787 IUPAC Name: 5-bromo-3-iodo-1H-pyridin-2-one SMILES: C1=C(C(=O)NC=C1Br)I