accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 381248-04-0

IUPAC Name: (2-chloropyridin-3-yl)boronic acid
Synonyms: 2-chloropyridin-3-ylboronic acid
Molecular Weight (g/mol): 157.36
Molecular Formula: C5H5BClNO2
InChi Key: VRDAOVQZVXYRNH-UHFFFAOYSA-N
SMILES: OB(O)C1=CC=CN=C1Cl
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Grade 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)