accessibility menu, dialog, popup

UserName

Viewing profile as user:
HClH2NNH2O

CAS RN 38148-63-9

IUPAC Name: hydrogen 4-(diaminomethylidene)cyclohexa-2,5-dien-1-one chloride
Synonyms: hydrogen 4-(diaminomethylidene)cyclohexa-2,5-dien-1-one chloride
Molecular Weight (g/mol): 172.61
Molecular Formula: C7H9ClN2O
InChi Key: OADOZRQXRQAJDT-UHFFFAOYSA-N
SMILES: [H+].[Cl-].NC(N)=C1C=CC(=O)C=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)