accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2NNHOONHNH2

CAS RN 3815-86-9

IUPAC Name: propanedihydrazide
Synonyms: propanedioic acid, dihydrazide
Molecular Weight (g/mol): 132.12
Molecular Formula: C3H8N4O2
InChi Key: PSIKPHJLTVSQFO-UHFFFAOYSA-N
SMILES: NNC(=O)CC(=O)NN

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results