accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 38167-72-5

IUPAC Name: 1-(2,6-diethylphenyl)-2,5-dihydro-1H-pyrrole-2,5-dione
Synonyms: 1-(2,6-diethylphenyl)pyrrole-2,5-dione
Molecular Weight (g/mol): 229.28
Molecular Formula: C14H15NO2
InChi Key: LNOKVKHZEYOLIQ-UHFFFAOYSA-N
SMILES: CCC1=CC=CC(CC)=C1N1C(=O)C=CC1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results