accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 381725-49-1

IUPAC Name: 6'-methoxy-2,3'-bipyridine
Synonyms: 6'-methoxy-2,3'-bipyridine
Molecular Weight (g/mol): 186.21
Molecular Formula: C11H10N2O
InChi Key: HRVJUCXZUHFRNP-UHFFFAOYSA-N
SMILES: COC1=NC=C(C=C1)C1=CC=CC=N1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results