accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 38185-56-7

IUPAC Name: 5-bromo-3-chloro-2-fluoropyridine
Synonyms: 5-bromo-3-chloro-2-fluoropyridine
Molecular Weight (g/mol): 210.43
Molecular Formula: C5H2BrClFN
InChi Key: WXKLHBHJIBAXQH-UHFFFAOYSA-N
SMILES: FC1=NC=C(Br)C=C1Cl
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results