accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3C(S)O(S)(S)O(R)H3C(S)O

CAS RN 382-45-6

IUPAC Name: (3aS,3bS,9aR,9bS,11aS)-9a,11a-dimethyl-1H,2H,3H,3aH,3bH,4H,5H,7H,8H,9H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthrene-1,7,10-trione
Synonyms: adrenosterone adrenosterone, 3h-labeled reichstein substance G reichstein's substance G adrenosterone, 3H-labeled 3h-labeled adrenosterone 3H-labeled adrenosterone
Molecular Weight (g/mol): 300.4
Molecular Formula: C19H24O3
InChi Key: RZRPTBIGEANTGU-IRIMSJTPSA-N
SMILES: C[C@]12CC(=O)[C@H]3[C@@H](CCC4=CC(=O)CC[C@]34C)[C@@H]1CCC2=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results