accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 38201-97-7

IUPAC Name: platinum(2+) tetraamine dihydroxide
Synonyms: platinum(2+) tetraamine dihydroxide
Molecular Weight (g/mol): 297.22
Molecular Formula: H14N4O2Pt
InChi Key: VSKCDODNDPOZKS-UHFFFAOYSA-L
SMILES: N.N.N.N.[OH-].[OH-].[Pt++]

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results