accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COCH3NOSOOCH3CH3H3C(E)

CAS RN 38202-27-6

IUPAC Name: (E)-(ethyl N-[(2,4,6-trimethylbenzenesulfonyl)oxy]ethanimidate)
Synonyms: (E)-(ethyl N-[(2,4,6-trimethylbenzenesulfonyl)oxy]ethanimidate)
Molecular Weight (g/mol): 285.36
Molecular Formula: C13H19NO4S
InChi Key: KQCBSWBQAXTILK-WYMLVPIESA-N
SMILES: CCO\C(C)=N\OS(=O)(=O)C1=C(C)C=C(C)C=C1C
Molecular Weight (g/mol) 
  • (5)
  • (1)
Percent Purity 
  • (1)
  • (2)
  • (3)
Quantity 
  • (2)
  • (2)
  • (2)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (3)
  • (2)
  • (5)
  • (1)
  • (1)
  • (2)
  • (3)
  • (2)
  • (2)
  • (2)
  • (3)