CAS RN 38215-38-2
IUPAC Name:
4,4'-diethynyl-1,1'-biphenyl
Synonyms:
4,4'-diethynyl-1,1'-biphenyl
Molecular Weight (g/mol):
202.26
Molecular Formula:
C16H10
InChi Key:
MXJJMQSKDPNPSX-UHFFFAOYSA-N
SMILES:
C#CC1=CC=C(C=C1)C1=CC=C(C=C1)C#C
1
–
1
of
1
results
1
4,4'-Diethynylbiphenyl 98.0+%, TCI America™
CAS: 38215-38-2 Molecular Formula: C16H10 Molecular Weight (g/mol): 202.26 MDL Number: MFCD00458964 InChI Key: MXJJMQSKDPNPSX-UHFFFAOYSA-N PubChem CID: 10013083 IUPAC Name: 4,4'-diethynyl-1,1'-biphenyl SMILES: C#CC1=CC=C(C=C1)C1=CC=C(C=C1)C#C