accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 38218-24-5

CH3CH3OCH3CH3OCH3CH3OIn

CAS RN 38218-24-5

IUPAC Name: indium(3+) tris(propan-2-olate)
Synonyms: indium(3+) tris(propan-2-olate)
Molecular Weight (g/mol): 292.08
Molecular Formula: C9H21InO3
InChi Key: OVZUSPADPSOQQN-UHFFFAOYSA-N
SMILES: [In+3].CC(C)[O-].CC(C)[O-].CC(C)[O-]
Percent Purity 
  • (2)
  • (3)
Physical Form 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)