accessibility menu, dialog, popup

UserName

Viewing profile as user:
BrOHO

CAS RN 38220-77-8

IUPAC Name: (5-bromo-1-benzofuran-2-yl)methanol
Synonyms: (5-bromo-1-benzofuran-2-yl)methanol
Molecular Weight (g/mol): 227.06
Molecular Formula: C9H7BrO2
InChi Key: JYYWIDBNICYLBN-UHFFFAOYSA-N
SMILES: OCC1=CC2=CC(Br)=CC=C2O1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results