accessibility menu, dialog, popup

UserName

Viewing profile as user:
FOHO

CAS RN 38226-10-7

IUPAC Name: 2-fluoro-6-hydroxybenzaldehyde
Synonyms: 2-fluoro-6-hydroxybenzaldehyde
Molecular Weight (g/mol): 140.11
Molecular Formula: C7H5FO2
InChi Key: FZIBGCDUHZBOLA-UHFFFAOYSA-N
SMILES: OC1=C(C=O)C(F)=CC=C1
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)