accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOOOOOOOOHoHo

CAS RN 38245-34-0

IUPAC Name: diholmium(3+) tricarbonate
Synonyms: diholmium(3+) tricarbonate
Molecular Weight (g/mol): 509.89
Molecular Formula: C3Ho2O9
InChi Key: FZKKGPOEHOOXQE-UHFFFAOYSA-H
SMILES: [Ho+3].[Ho+3].[O-]C([O-])=O.[O-]C([O-])=O.[O-]C([O-])=O
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results