accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 38245-38-4

IUPAC Name: dineodymium(3+) tricarbonate
Synonyms: dineodymium(3+) tricarbonate
Molecular Weight (g/mol): 468.51
Molecular Formula: C3Nd2O9
InChi Key: UTWHRPIUNFLOBE-UHFFFAOYSA-H
SMILES: [Nd+3].[Nd+3].[O-]C([O-])=O.[O-]C([O-])=O.[O-]C([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results