accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 38245-39-5

IUPAC Name: diyttrium(3+) tricarbonate
Synonyms: diyttrium(3+) tricarbonate
Molecular Weight (g/mol): 357.84
Molecular Formula: C3O9Y2
InChi Key: QVOIJBIQBYRBCF-UHFFFAOYSA-H
SMILES: [Y+3].[Y+3].[O-]C([O-])=O.[O-]C([O-])=O.[O-]C([O-])=O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (2)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)