accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NHOOCH3CH3H3C

CAS RN 38267-76-4

IUPAC Name: tert-butyl N-ethylcarbamate
Synonyms: tert-butyl N-ethylcarbamate
Molecular Weight (g/mol): 145.2
Molecular Formula: C7H15NO2
InChi Key: ISTGQSQWSKCNFJ-UHFFFAOYSA-N
SMILES: CCNC(=O)OC(C)(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)