accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 383134-85-8

IUPAC Name: 2-(5-fluoro-2-methoxyphenyl)acetic acid
Synonyms: (5-fluoro-2-methoxyphenyl)acetic acid
Molecular Weight (g/mol): 184.17
Molecular Formula: C9H9FO3
InChi Key: WGZCWKASVDIAOH-UHFFFAOYSA-N
SMILES: COC1=C(CC(O)=O)C=C(F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results