accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 383177-55-7

IUPAC Name: 4,4,5,5,6,6,7,7,8,8,8-undecafluorooctan-3-one
Synonyms: 4,4,5,5,6,6,7,7,8,8,8-undecafluorooctan-3-one
Molecular Weight (g/mol): 326.11
Molecular Formula: C8H5F11O
InChi Key: VHTNJYGNFUMJTN-UHFFFAOYSA-N
SMILES: CCC(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results