accessibility menu, dialog, popup

UserName

Viewing profile as user:
NNOOHNH(E/Z)

CAS RN 38369-95-8

IUPAC Name: 4-(4,6-diphenyl-1,2-dihydro-1,3,5-triazin-2-ylidene)-3-hydroxycyclohexa-2,5-dien-1-one
Synonyms: 4-(4,6-diphenyl-1H-1,3,5-triazin-2-ylidene)-3-hydroxycyclohexa-2,5-dien-1-one
Molecular Weight (g/mol): 341.37
Molecular Formula: C21H15N3O2
InChi Key: JUMDJIDYKLZVAF-UHFFFAOYSA-N
SMILES: OC1=CC(=O)C=CC1=C1NC(=NC(=N1)C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results