accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 38396-39-3

IUPAC Name: 1-butyl-N-(2,6-dimethylphenyl)piperidine-2-carboxamide
Synonyms: bupivacaine carbostesin transdur-bupivacaine dolanaest DL-bupivacaine svedocain sin vasoconstr carbonate, bupivacaine sensorcaine-mpf spinal bupivacainum bupivan
Molecular Weight (g/mol): 288.44
Molecular Formula: C18H28N2O
InChi Key: LEBVLXFERQHONN-UHFFFAOYNA-N
SMILES: CCCCN1CCCCC1C(=O)NC1=C(C)C=CC=C1C
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)