accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 38422-62-7

IUPAC Name: 5-tert-butyl-2-methylfuran-3-carboxylate
Synonyms: 5-tert-butyl-2-methylfuran-3-carboxylate
Molecular Weight (g/mol): 181.21
Molecular Formula: C10H13O3
InChi Key: NWWAPYOVUAEIJB-UHFFFAOYSA-M
SMILES: CC1=C(C=C(O1)C(C)(C)C)C([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results