accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOO

CAS RN 38425-65-9

IUPAC Name: 2-(2,3-dihydro-1H-inden-1-yl)acetic acid
Synonyms: 2,3-dihydro-1H-inden-1-ylacetic acid
Molecular Weight (g/mol): 176.22
Molecular Formula: C11H12O2
InChi Key: RJVZEPWRJJBXLH-UHFFFAOYNA-N
SMILES: OC(=O)CC1CCC2=CC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results